missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalaninol, 95%

CAS: 16088-07-6 | C9H13NO | 151.209 g/mol

$78.03 - $1083.90

Chemical Identifiers

CAS 16088-07-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00066689
InChI Key STVVMTBJNDTZBF-UHFFFAOYSA-N
Synonym dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol
PubChem CID 76652
IUPAC Name 2-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CC(CO)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6356203
View Documents
Thermo Scientific Chemicals
H6356203
1 g
Each for $78.03
Only null left
 
AAH6356206
View Documents
Thermo Scientific Chemicals
H6356206
5 g
Each for $275.38
Only null left
 
AAH6356214
View Documents
Thermo Scientific Chemicals
H6356214
25 g
Each for $1,083.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16088-07-6
151.209
STVVMTBJNDTZBF-UHFFFAOYSA-N
76652
C1=CC=C(C=C1)CC(CO)N
C9H13NO
MFCD00066689
dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol
2-amino-3-phenylpropan-1-ol

Specifications

69°C to 73°C
C9H13NO
UN3259
dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol
C1=CC=C(C=C1)CC(CO)N
1 g
76652
96%
DL-Phenylalaninol
16088-07-6
MFCD00066689
2208237
STVVMTBJNDTZBF-UHFFFAOYSA-N
2-amino-3-phenylpropan-1-ol
151.209
151.21
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only