Learn More
DL-Phenylalaninol, 95%
CAS: 16088-07-6 | C9H13NO | 151.209 g/mol
Supplier: Thermo Scientific Chemicals H6356203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 69°C to 73°C | |
| 16088-07-6 | |
| MFCD00066689 | |
| 2208237 | |
| STVVMTBJNDTZBF-UHFFFAOYSA-N | |
| 2-amino-3-phenylpropan-1-ol | |
| 151.209 | |
| 151.21 | |
| Solid |
| DL-Phenylalaninol | |
| C9H13NO | |
| UN3259 | |
| dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol | |
| C1=CC=C(C=C1)CC(CO)N | |
| 1 g | |
| 76652 | |
| 96% |
Chemical Identifiers
| 16088-07-6 | |
| 151.209 | |
| STVVMTBJNDTZBF-UHFFFAOYSA-N | |
| 76652 | |
| C1=CC=C(C=C1)CC(CO)N |
| C9H13NO | |
| MFCD00066689 | |
| dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol | |
| 2-amino-3-phenylpropan-1-ol |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only