missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalanine methyl ester hydrochloride, 98%

CAS: 5619-07-8 | C10H14ClNO2 | 215.68 g/mol

Supplier:  Thermo Scientific Chemicals A1275814

 View more versions of this product

Catalog No. AAA1275814


Only null left

Description

Description

DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
Specifications

Specifications

∼160°C (decomposition)
5619-07-8
MFCD00066113
dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-phenylpropanoate hydrochloride, h-dl-phe-ome.hcl, h-l-phe-ome hcl, phenylalanine methyl ester hydrochloride, l-phenylalanine methylester hydrochloride, l-methyl phenylalanate hydrochloride, s-phenylalanine methyl ester hydrochloride, acmc-209oxm, acmc-1bga3
SWVMLNPDTIFDDY-UHFFFAOYNA-N
methyl 2-amino-3-phenylpropanoate;hydrochloride
215.68
215.68
DL-Phenylalanine methyl ester hydrochloride
C10H14ClNO2
3597947
Slightly soluble in water.
Cl.COC(=O)C(N)CC1=CC=CC=C1
25 g
3084017
98%

Chemical Identifiers

5619-07-8
215.68
SWVMLNPDTIFDDY-UHFFFAOYNA-N
3084017
Cl.COC(=O)C(N)CC1=CC=CC=C1
C10H14ClNO2
MFCD00066113
dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-phenylpropanoate hydrochloride, h-dl-phe-ome.hcl, h-l-phe-ome hcl, phenylalanine methyl ester hydrochloride, l-phenylalanine methylester hydrochloride, l-methyl phenylalanate hydrochloride, s-phenylalanine methyl ester hydrochloride, acmc-209oxm, acmc-1bga3
methyl 2-amino-3-phenylpropanoate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 227-049-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only