missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalanine methyl ester hydrochloride, 98%

Catalog No. p-7020710
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1275806 5 g
AAA1275814 25 g
AAA1275822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1275806 Supplier Thermo Scientific Chemicals Supplier No. A1275806

CAS: 5619-07-8 | C10H14ClNO2 | 215.68 g/mol

DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine methyl ester hydrochloride, is an aminoacid which is widely used in pharmaceuticals and foods.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 5619-07-8
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00066113
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
Synonym dl-phenylalanine methyl ester hydrochloride, methyl 2-amino-3-phenylpropanoate hydrochloride, h-dl-phe-ome.hcl, h-l-phe-ome hcl, phenylalanine methyl ester hydrochloride, l-phenylalanine methylester hydrochloride, l-methyl phenylalanate hydrochloride, s-phenylalanine methyl ester hydrochloride, acmc-209oxm, acmc-1bga3
PubChem CID 3084017
IUPAC Name methyl 2-amino-3-phenylpropanoate;hydrochloride
SMILES Cl.COC(=O)C(N)CC1=CC=CC=C1

Specifications

Melting Point ∼160°C (decomposition)
Beilstein 3597947
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 215.68
Percent Purity 98%
Chemical Name or Material DL-Phenylalanine methyl ester hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 227-049-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only