missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Phenylalanine, 99%
DL-Phenylalanine, 99%, C9H11NO2, CAS Number-150-30-1, 2-amino-3-phenylpropionic acid, alanine, phenyl-, dl, dl-2-amino-3-phenylpropanoic acid, dl-3-phenylalanine, dlpa, dl-phenylalanine, h-dl-phe-oh, phenylalanin, phenylalanine, phenylalanine, dl, 100g, 14,498, CHEBI:28044, White, 205-756-7, 165.19 | CAS: 150-30-1 | C9H11NO2 | 165.19 g/mol
$73.82 - $125.83
Chemical Identifiers
| CAS | 150-30-1 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.19 |
| MDL Number | MFCD00064225 |
| InChI Key | COLNVLDHVKWLRT-UHFFFAOYNA-N |
| Synonym | dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl |
| PubChem CID | 994 |
| ChEBI | CHEBI:28044 |
| IUPAC Name | 2-amino-3-phenylpropanoic acid |
| SMILES | NC(CC1=CC=CC=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC164570250
|
Thermo Scientific Chemicals
164570250 |
25 g | Plastic bottle |
Each for $73.82
|
|
||||
|
AC164571000
|
Thermo Scientific Chemicals
164571000 |
100 g | Plastic bottle |
Each for $125.83
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 150-30-1 | |
| 165.19 | |
| COLNVLDHVKWLRT-UHFFFAOYNA-N | |
| 994 | |
| 2-amino-3-phenylpropanoic acid |
| C9H11NO2 | |
| MFCD00064225 | |
| dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl | |
| CHEBI:28044 | |
| NC(CC1=CC=CC=C1)C(O)=O |
Specifications
| 150-30-1 | |
| White | |
| Authentic | |
| Plastic bottle | |
| C6H5CH2CH(NH2)CO2H | |
| 25 g | |
| 15,7382 | |
| Solubility in water: 14.11g/L (25°C) | |
| NC(CC1=CC=CC=C1)C(O)=O | |
| 165.19 | |
| CHEBI:28044 | |
| 99% | |
| DL-Phenylalanine |
| 266.0°C | |
| 0.3% (105°C, 2 hrs) max. | |
| 99% | |
| C9H11NO2 | |
| MFCD00064225 | |
| 14,498 | |
| dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl | |
| COLNVLDHVKWLRT-UHFFFAOYNA-N | |
| 2-amino-3-phenylpropanoic acid | |
| 994 | |
| 165.19 | |
| Powder |
Safety and Handling
EINECSNumber : 205-756-7
RUO – Research Use Only