missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Phenylalanine, 99%

Catalog No. p-7080552
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1013214 25 g
AAA1013222 100 g
AAA1013236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1013214 Supplier Thermo Scientific Chemicals Supplier No. A1013214

CAS: 150-30-1 | C9H11NO2 | 165.19 g/mol

DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

Solubility
Soluble in water. Insoluble in ethyl ether, benzene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 150-30-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064225
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
PubChem CID 994
ChEBI CHEBI:28044
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∼266°C (decomposition)
Beilstein 1910407
Merck Index 14,7271
Solubility Information Soluble in water. Insoluble in ethyl ether,benzene.
Quantity 25 g
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material DL-Phenylalanine
EINECSNumber 205-756-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only