missing translation for 'onlineSavingsMsg'
Learn More

DL-Norvaline, ≥98.5% (HPLC); 98.5-101.5% (NT), MilliporeSigma™ Supelco™

Catalog No. 111015448
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015448 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015448 Supplier Merck Emd Millipore Supplier No. 53721100MG
Only null left
Food and Beverages

Specifications

CAS 760-78-1
Linear Formula CH3 CH2 CH2 CH(NH2)COOH
MDL Number MFCD00064420
Synonym (+/-)-2-Aminopentanoic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 117.15
Percent Purity ≥98.5% (HPLC); 98.5 to 101.5% (NT)
Grade Analytical Standard