missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+%
CAS: 62697-73-8 | C5H11NO3S | 165.21 g/mol
$126.22 - $466.04
Chemical Identifiers
| CAS | 62697-73-8 |
|---|---|
| Molecular Formula | C5H11NO3S |
| Molecular Weight (g/mol) | 165.21 |
| MDL Number | MFCD00002620 |
| InChI Key | QEFRNWWLZKMPFJ-UHFFFAOYNA-N |
| Synonym | dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl |
| PubChem CID | 847 |
| ChEBI | CHEBI:49033 |
| SMILES | CS(=O)CCC(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1808106
|
Thermo Scientific Chemicals
A1808106 |
5 g |
Each for $126.22
|
|
|||||
|
AAA1808114
|
Thermo Scientific Chemicals
A1808114 |
25 g |
Each for $466.04
|
|
|||||
Description
DL-Methionine sulfoxide is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Methionine sulfoxide is used as pharmaceutical intermediates.
Solubility
Soluble in water and Hcl.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
Chemical Identifiers
| 62697-73-8 | |
| 165.21 | |
| QEFRNWWLZKMPFJ-UHFFFAOYNA-N | |
| 847 | |
| CS(=O)CCC(N)C(O)=O |
| C5H11NO3S | |
| MFCD00002620 | |
| dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl | |
| CHEBI:49033 |
Specifications
| ∼230°C (decomposition) | |
| C5H11NO3S | |
| dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl | |
| QEFRNWWLZKMPFJ-UHFFFAOYNA-N | |
| 2-amino-4-methylsulfinylbutanoic acid | |
| 165.21 | |
| CHEBI:49033 | |
| ≥98% |
| 62697-73-8 | |
| MFCD00002620 | |
| Soluble in water and Hcl. | |
| CS(=O)CCC(N)C(O)=O | |
| 5 g | |
| 847 | |
| 165.21 | |
| DL-Methionine sulfoxide |
Safety and Handling
EINECSNumber : 263-700-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only