missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Methionine sulfoxide, 98+%

CAS: 62697-73-8 | C5H11NO3S | 165.21 g/mol

Supplier:  Thermo Scientific Chemicals A1808114

 View more versions of this product

Catalog No. AAA1808114

Only null left
Explore available promotions

Description

Description

DL-Methionine sulfoxide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Methionine sulfoxide is used as pharmaceutical intermediates.

Solubility
Soluble in water and Hcl.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container
Specifications

Specifications

∼230°C (decomposition)
62697-73-8
MFCD00002620
Soluble in water and Hcl.
CS(=O)CCC(N)C(O)=O
25 g
847
165.21
DL-Methionine sulfoxide
C5H11NO3S
dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl
QEFRNWWLZKMPFJ-UHFFFAOYNA-N
2-amino-4-methanesulfinylbutanoic acid
165.21
CHEBI:49033
≥98%

Chemical Identifiers

62697-73-8
165.21
QEFRNWWLZKMPFJ-UHFFFAOYNA-N
847
CS(=O)CCC(N)C(O)=O
C5H11NO3S
MFCD00002620
dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl
CHEBI:49033
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 263-700-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only