missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Mercaptosuccinic acid, 99%
CAS: 70-49-5 | C4H6O4S | 150.15 g/mol
$111.15 - $111.15
Chemical Identifiers
| CAS | 70-49-5 |
|---|---|
| Molecular Formula | C4H6O4S |
| Molecular Weight (g/mol) | 150.15 |
| InChI Key | NJRXVEJTAYWCQJ-UHFFFAOYSA-N |
| Synonym | mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid |
| PubChem CID | 6268 |
| ChEBI | CHEBI:38705 |
| IUPAC Name | 2-sulfanylbutanedioic acid |
| SMILES | C(C(C(=O)O)S)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC125551000
|
Thermo Scientific Chemicals
125551000 |
100 g | Glass bottle |
Each for $111.15
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 70-49-5 | |
| 150.15 | |
| mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid | |
| CHEBI:38705 | |
| C(C(C(=O)O)S)C(=O)O |
| C4H6O4S | |
| NJRXVEJTAYWCQJ-UHFFFAOYSA-N | |
| 6268 | |
| 2-sulfanylbutanedioic acid |
Specifications
| 70-49-5 | |
| White | |
| 99% | |
| C4H6O4S | |
| 100 g | |
| 15,9496 | |
| Solubility in water: 150g/L (20°C). Other solubilities: 500g/L ethanol (20°C),soluble in acetone,moderately soluble in ether,chloroform,practically insoluble in benzene | |
| C(C(C(=O)O)S)C(=O)O | |
| 150.15 | |
| CHEBI:38705 | |
| 99% | |
| DL-Mercaptosuccinic acid |
| 152°C to 155°C | |
| Authentic | |
| Glass bottle | |
| HO2CCH2CH(SH)CO2H | |
| 03,439 | |
| mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid | |
| NJRXVEJTAYWCQJ-UHFFFAOYSA-N | |
| 2-sulfanylbutanedioic acid | |
| 6268 | |
| 150.15 | |
| Crystalline Powder |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 200-736-4
RUO – Research Use Only