missing translation for 'onlineSavingsMsg'
Learn More

DL-Menthol, Crystal, FCC, Spectrum™ Chemical

M1776, 89-78-1, C10H20O

Supplier:  Spectrum Chemical Mfg Cor M1776500GM

Quantity 500 g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18602291


Only null left

Description

Description

Spectrum™ Chemical DL-Menthol, Crystal, FCC is used as a flavor and fragrance additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

89-78-1
156.27
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
C10H20O
NOOLISFMXDJSKH-KXUCPTDWSA-N
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

DL-Menthol
100%
31°C to 44°C
C10H20O
0.05%
NOOLISFMXDJSKH-KXUCPTDWSA-N
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
FCC
89-78-1
212°C
Amber Glass Bottle
500 g
-2° to +2°
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
156.27
Low Melting Crystals or Crystalline Mass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions