missing translation for 'onlineSavingsMsg'
Learn More

DL-Menthol, Crystal, FCC, Spectrum™ Chemical

Catalog No. p-7157767
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602290 100 g Amber Glass Bottle
18602291 500 g Amber Glass Bottle
18602292 2.5 kg Amber Glass Bottle
18602293 10 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602290 Supplier Spectrum Chemical Mfg Cor Supplier No. M1776100GM
Only null left

M1776, 89-78-1, C10H20O

Spectrum™ Chemical DL-Menthol, Crystal, FCC is used as a flavor and fragrance additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
IUPAC Name (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

CAS Max % 100%
Boiling Point 212°C
Melting Range 31°C to 44°C
Packaging Amber Glass Bottle
Quantity 100 g
Residue after Evaporation 0.05%
Specific Rotation -2° to +2°
Grade FCC
Physical Form Low Melting Crystals or Crystalline Mass
Chemical Name or Material DL-Menthol