missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Leucylglycylglycine 99.0+%, TCI America™
$29.83 - $145.40
Specifications
| Purity | ≥99.0% (T) |
|---|---|
| CAS | 4337-37-5 |
| Color | White-Yellow |
| Format | Crystal-Powder at 20°C |
| Molecular Weight (g/mol) | 245.28 |
Specifications
| ≥99.0% (T) | |
| White-Yellow | |
| 245.28 |
| 4337-37-5 | |
| Crystal-Powder at 20°C | |
| DL-Leucylglycylglycine |