accessibility menu, dialog, popup

UserName

CAS RN 4337-37-5

H3CCH3NH2OHNONHOHO

CAS RN 4337-37-5

IUPAC Name: 2-[2-(2-amino-4-methylpentanamido)acetamido]acetic acid
Synonyms: glycine, N-(N-L-leucylglycyl)-
Molecular Weight (g/mol): 245.28
Molecular Formula: C10H19N3O4
InChi Key: VWHGTYCRDRBSFI-UHFFFAOYNA-N
SMILES: CC(C)CC(N)C(=O)NCC(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results