missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactide 98.0+%, TCI America™

Supplier:  TCI America L0091100G

 View more versions of this product

Catalog No. L0091100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

DL-Lactide
95-96-5
142°C
Crystal-Powder at 20°C
MFCD00011685,MFCD00082566
JJTUDXZGHPGLLC-UHFFFAOYNA-N
3,6-dimethyl-1,4-dioxane-2,5-dione
7272
≥98.0% (T)
124°C
White
100 g
C6H8O4
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
CC1OC(=O)C(C)OC1=O
144.13
144.13

Chemical Identifiers

95-96-5
144.13
JJTUDXZGHPGLLC-UHFFFAOYNA-N
7272
CC1OC(=O)C(C)OC1=O
C6H8O4
MFCD00011685,MFCD00082566
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
3,6-dimethyl-1,4-dioxane-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5295

RTECSNumber : JH1100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions