missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactide 98.0+%, TCI America™

Catalog No. p-7131818
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L009125G 25 g
L0091100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L009125G Supplier TCI America Supplier No. L009125G
Only null left

Chemical Identifiers

CAS 95-96-5
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00011685,MFCD00082566
InChI Key JJTUDXZGHPGLLC-UHFFFAOYNA-N
Synonym dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
PubChem CID 7272
IUPAC Name 3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC1OC(=O)C(C)OC1=O

Specifications

Melting Point 124°C
Color White
Boiling Point 142°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 144.13
Percent Purity ≥98.0% (T)
Chemical Name or Material DL-Lactide
EINECSNumber (5)-5295
RTECSNumber JH1100000
TSCA Yes