missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactide, 99%

CAS: 95-96-5 | C6H8O4 | 144.13 g/mol

Supplier:  Thermo Scientific Chemicals L0902618

 View more versions of this product

Catalog No. AAL0902618


Only null left
Explore available promotions

Description

Description

DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.

Solubility
Soluble in chloroform and methanol. Slightly soluble in benzene.

Notes
Moisture senstive. Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.
Specifications

Specifications

DL-Lactide
95-96-5
50 g
MFCD00011685,MFCD00082566
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
JJTUDXZGHPGLLC-UHFFFAOYNA-N
3,6-dimethyl-1,4-dioxane-2,5-dione
7272
99%
122°C to 126°C
138°C to 142°C (8 mmHg)
C6H8O4
82121
Soluble in chloroform and methanol. Slightly soluble in benzene.
CC1OC(=O)C(C)OC1=O
144.13
144.13

Chemical Identifiers

95-96-5
144.13
JJTUDXZGHPGLLC-UHFFFAOYNA-N
7272
CC1OC(=O)C(C)OC1=O
C6H8O4
MFCD00011685,MFCD00082566
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
3,6-dimethyl-1,4-dioxane-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-468-3

RTECSNumber : JH1100000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only