missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactide, 99%

Catalog No. p-7044994
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0902609 10 g
AAL0902618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0902609 Supplier Thermo Scientific Chemicals Supplier No. L0902609
Only null left

CAS: 95-96-5 | C6H8O4 | 144.13 g/mol

DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.

Solubility
Soluble in chloroform and methanol. Slightly soluble in benzene.

Notes
Moisture senstive. Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.

Chemical Identifiers

CAS 95-96-5
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00011685,MFCD00082566
InChI Key JJTUDXZGHPGLLC-UHFFFAOYNA-N
Synonym dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
PubChem CID 7272
IUPAC Name 3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC1OC(=O)C(C)OC1=O

Specifications

Melting Point 122°C to 126°C
Boiling Point 138°C to 142°C (8 mmHg)
Quantity 10 g
Beilstein 82121
Solubility Information Soluble in chloroform and methanol. Slightly soluble in benzene.
Formula Weight 144.13
Percent Purity 99%
Chemical Name or Material DL-Lactide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-468-3
RTECSNumber JH1100000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only