missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactic acid, 85+%, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC412965000
Change view
Click to view available options
Quantity:
500 g
Packaging:
EcoSafPak™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412965000 500 g EcoSafPak™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412965000 Supplier Thermo Scientific Chemicals Supplier No. 412965000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-21-5 | C3H6O3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Color Colorless
Density 1.2000g/mL
Quantity 500 g
Packaging EcoSafPak™ Glass Bottle
Physical Form Slightly Viscous Liquid
Residue after Ignition 0.02% max.
Assay Percent Range 85+%
Linear Formula CH3CH(OH)CO2H
Merck Index 15, 5384
Formula Weight 90.08
Percent Purity 85.0 to 90.0%
Grade ACS Reagent
Chemical Name or Material DL-Lactic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-018-0
RTECSNumber OD2800000
TSCA TSCA

RUO – Research Use Only