missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoserine 95.0+%, TCI America™

Catalog No. p-7131324
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0309100MG 100 mg
I03091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0309100MG Supplier TCI America Supplier No. I0309100MG
Only null left

Chemical Identifiers

CAS 565-71-9
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00008138
InChI Key BMYNFMYTOJXKLE-UHFFFAOYSA-N
Synonym dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947
PubChem CID 11267
IUPAC Name 3-amino-2-hydroxypropanoic acid
SMILES C(C(C(=O)O)O)N

Specifications

Melting Point 235°C
Color White
Quantity 100 mg
Formula Weight 105.09
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Isoserine