accessibility menu, dialog, popup

UserName

CAS RN 565-71-9

NH2OHOHO

CAS RN 565-71-9

IUPAC Name: 3-amino-2-hydroxypropanoic acid
Synonyms: isoserine
Molecular Weight (g/mol): 105.09
Molecular Formula: C3H7NO3
InChi Key: BMYNFMYTOJXKLE-UHFFFAOYNA-N
SMILES: NCC(O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)