missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Isoserine 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 235°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 105.09 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-Isoserine |
Safety and Handling
| TSCA | No |