missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Isoleucine (mixture of 4 stereoisomers) 98.0+%, TCI America™
$80.85 - $275.30
Chemical Identifiers
| CAS | 443-79-8 |
|---|---|
| Molecular Formula | C6H13NO2 |
| Molecular Weight (g/mol) | 131.18 |
| MDL Number | MFCD00004268 |
| InChI Key | AGPKZVBTJJNPAG-WHFBIAKZSA-N |
| Synonym | d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh |
| PubChem CID | 94206 |
| ChEBI | CHEBI:20899 |
| IUPAC Name | DL-2-amino-3-methylpentanoic acid |
| SMILES | CC[C@H](C)[C@H](N)C(O)=O |
Chemical Identifiers
| 443-79-8 | |
| 131.18 | |
| AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
| 94206 | |
| DL-2-amino-3-methylpentanoic acid |
| C6H13NO2 | |
| MFCD00004268 | |
| d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh | |
| CHEBI:20899 | |
| CC[C@H](C)[C@H](N)C(O)=O |
Specifications
| 443-79-8 | |
| C6H13NO2 | |
| d-alloisoleucine, d-allo-isoleucine, 2r,3s-2-amino-3-methylpentanoic acid, allo-d-isoleucine, alloisoleucine, d, h-d-allo-ile-oh, threo-d-isoleucine, dl-allo-isoleucine, hile, h-dl-allo-ile-oh | |
| CC[C@H](C)[C@H](N)C(O)=O | |
| 5 g | |
| 94206 | |
| 131.18 | |
| Crystalline Powder |
| White | |
| MFCD00004268 | |
| AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
| DL-2-amino-3-methylpentanoic acid | |
| 131.18 | |
| CHEBI:20899 | |
| ≥98.0% (T) | |
| DL-Isoleucine (mixture of 4 stereoisomers) |
Safety and Handling
EINECSNumber : (9)-1560
TSCA : Yes