accessibility menu, dialog, popup

UserName

CAS RN 443-79-8

H3C(S)CH3(S)NH2OHO

CAS RN 443-79-8

IUPAC Name: DL-2-amino-3-methylpentanoic acid
Synonyms: Ile|isoleucina|2-amino-3-methylvaleric acid|isoleucinum|erythro-L-isoleucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N
SMILES: CC[C@H](C)[C@H](N)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)