missing translation for 'onlineSavingsMsg'
Learn More

DL-Isoborneol, 93%, tech.

Catalog No. p-150229
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122490250 25 g Plastic bottle
AC122495000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122490250 Supplier Thermo Scientific Chemicals Supplier No. 122490250
Only null left

CAS: 124-76-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00074821
InChI Key DTGKSKDOIYIVQL-SZBHIRRCSA-N
Synonym dl-isoborneol
PubChem CID 126961757
IUPAC Name (1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C2CCC1(C(C2)O)C)C

Specifications

Melting Point 208.0°C to 214.0°C
Color Yellow
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 06, II, 80
Merck Index 15, 5173
Solubility Information Solubility in water: insoluble in water. Other solubilities: readily soluble in alcohol,chloroform and ether
Formula Weight 154.25
Percent Purity 93%
Grade Technical
Physical Form Powder
Chemical Name or Material DL-Isoborneol, tech., 93%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 204-712-4
RTECSNumber NP7300000
TSCA TSCA

RUO – Research Use Only