missing translation for 'onlineSavingsMsg'
Learn More

DL-Homophenylalanine 97.0+%, TCI America™

Catalog No. H13295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13295G 5 g
H13291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13295G Supplier TCI America Supplier No. H13295G
Only null left

Chemical Identifiers

CAS 1012-05-1
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00063091,MFCD00063092
InChI Key JTTHKOPSMAVJFE-UHFFFAOYNA-N
Synonym 2-Amino-4-phenylbutyric Acid
PubChem CID 102530
IUPAC Name 2-amino-4-phenylbutanoic acid
SMILES NC(CCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 294°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥97.0% (HPLC,T)
Chemical Name or Material DL-Homophenylalanine