accessibility menu, dialog, popup

UserName

OOHNH2

CAS RN 1012-05-1

IUPAC Name: 2-amino-4-phenylbutanoic acid
Synonyms: homophenylalanine
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: JTTHKOPSMAVJFE-UHFFFAOYNA-N
SMILES: NC(CCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results