missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DL-Homocysteinethiolactone hydrochloride, 99%

$69.08 - $200.18

Chemical Identifiers

CAS 6038-19-3
Molecular Formula C4H7NOS·HCl
Molecular Weight (g/mol) 153.63
MDL Number MFCD00012724
InChI Key ZSEGSUBKDDEALH-UHFFFAOYSA-N
Synonym dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
PubChem CID 110753
IUPAC Name 3-aminothiolan-2-one;hydrochloride
SMILES C1CSC(=O)C1N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC120830250
View Documents Promotion Details
Thermo Scientific™
120830250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC120831000
View Documents Promotion Details
Thermo Scientific™
120831000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6038-19-3
153.63
ZSEGSUBKDDEALH-UHFFFAOYSA-N
110753
C1CSC(=O)C1N.Cl
C4H7NOS·HCl
MFCD00012724
dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
3-aminothiolan-2-one;hydrochloride

Specifications

6038-19-3
99%
MFCD00012724
dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
>140°C
C1CSC(=O)C1N.Cl
153.63
153.63
DL-Homocysteinethiolactone hydrochloride, 99%
Authentic
C4H7NOS·HCl
15, 4772
Solubility in water: 740.5g/L (20 c)
ZSEGSUBKDDEALH-UHFFFAOYSA-N
3-aminothiolan-2-one;hydrochloride
110753
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-923-3

RTECSNumber : XN1928400

SDS
Documents

Documents

RUO – Research Use Only