missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Homocysteinethiolactone hydrochloride, 99%

Catalog No. AC120830250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120830250 25 g Glass bottle
AC120831000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC120830250 Supplier Thermo Scientific Chemicals Supplier No. 120830250

CAS: 6038-19-3 | C4H7NOS·HCl | 153.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6038-19-3
MDL Number MFCD00012724
InChI Key ZSEGSUBKDDEALH-UHFFFAOYSA-N
Synonym dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
PubChem CID 110753
IUPAC Name 3-aminothiolan-2-one;hydrochloride
SMILES C1CSC(=O)C1N.Cl

Specifications

Melting Point 199.0°C to 203.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H7NOS·HCl
Quantity 25 g
Merck Index 15, 4772
Solubility Information Solubility in water: 740.5g/L (20 c)
Sublimation Point >140°C
Molecular Weight (g/mol) 153.63
Formula Weight 153.63
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Homocysteinethiolactone hydrochloride, 99%
Show More Show Less
EINECSNumber 227-923-3
RTECSNumber XN1928400

RUO – Research Use Only