missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocysteine 90.0+%, TCI America™

Catalog No. H01591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01591G 1 g
H01595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01591G Supplier TCI America Supplier No. H01591G

Chemical Identifiers

CAS 454-29-5
Molecular Formula C4H9NO2S
Molecular Weight (g/mol) 135.181
MDL Number MFCD00004898
InChI Key FFFHZYDWPBMWHY-UHFFFAOYSA-N
PubChem CID 778
ChEBI CHEBI:17230
IUPAC Name 2-amino-4-sulfanylbutanoic acid
SMILES C(CS)C(C(=O)O)N

Specifications

Melting Point 233°C
Color White
Quantity 1 g
Formula Weight 135.18
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Homocysteine
RTECSNumber MT0175000
TSCA No