CAS RN 454-29-5
CAS RN 454-29-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DL-Homocysteine 90.0+%, TCI America™
CAS: 454-29-5 Molecular Formula: C4H9NO2S Molecular Weight (g/mol): 135.181 MDL Number: MFCD00004898 InChI Key: FFFHZYDWPBMWHY-UHFFFAOYSA-N PubChem CID: 778 ChEBI: CHEBI:17230 IUPAC Name: 2-amino-4-sulfanylbutanoic acid SMILES: C(CS)C(C(=O)O)N