missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Glutamic acid monohydrate, 98+%
CAS: 19285-83-7 | C5H11NO5 | 165.145 g/mol
$57.73 - $398.96
Chemical Identifiers
| CAS | 19285-83-7 |
|---|---|
| Molecular Formula | C5H11NO5 |
| Molecular Weight (g/mol) | 165.145 |
| MDL Number | MFCD00150703 |
| InChI Key | OZDAOHVKBFBBMZ-UHFFFAOYSA-N |
| Synonym | dl-glutamic acid monohydrate, a-aminoglutarate, dl-glutamic acid hydrate, dl-glutamic acid hydrate, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, dl-2-aminopentanedioic acid, 2-aminopentanedioic acid, oxamethane, hydrate, hydrate |
| PubChem CID | 16219385 |
| IUPAC Name | 2-aminopentanedioic acid;hydrate |
| SMILES | C(CC(=O)O)C(C(=O)O)N.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1771914
|
Thermo Scientific Chemicals
A1771914 |
25 g |
Each for $57.73
|
|
|||||
|
AAA1771922
|
Thermo Scientific Chemicals
A1771922 |
100 g |
Each for $127.27
|
|
|||||
|
AAA1771936
|
Thermo Scientific Chemicals
A1771936 |
500 g |
Each for $398.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19285-83-7 | |
| 165.145 | |
| OZDAOHVKBFBBMZ-UHFFFAOYSA-N | |
| 16219385 | |
| C(CC(=O)O)C(C(=O)O)N.O |
| C5H11NO5 | |
| MFCD00150703 | |
| dl-glutamic acid monohydrate, a-aminoglutarate, dl-glutamic acid hydrate, dl-glutamic acid hydrate, glutaminsaeure hydrate, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, dl-2-aminopentanedioic acid, 2-aminopentanedioic acid, oxamethane, hydrate, hydrate | |
| 2-aminopentanedioic acid;hydrate |
Specifications
| ∼200°C (decomposition) | |
| C5H11NO5 | |
| 4155097 | |
| dl-glutamic acid monohydrate, a-aminoglutarate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, dl-2-aminopentanedioic acid, dl-2-aminopentanedioic acid, 2-aminopentanedioic acid, oxamethane, hydrate, hydrate | |
| C(CC(=O)O)C(C(=O)O)N.O | |
| 2-aminopentanedioic acid;hydrate | |
| 165.145 | |
| 165.15 (147.13 Anhydrous) | |
| DL-Glutamic acid monohydrate |
| 19285-83-7 | |
| MFCD00150703 | |
| 14,4469 | |
| OZDAOHVKBFBBMZ-UHFFFAOYSA-N | |
| 1.46 | |
| 25 g | |
| 16219385 | |
| 99% |
Safety and Handling
EINECSNumber : 210-522-2
RTECSNumber : LZ9648000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only