Learn More
Thermo Scientific Chemicals DL-Glutamic acid monohydrate, 98+%
CAS: 19285-83-7 | C5H11NO5 | 165.145 g/mol
Supplier: Thermo Scientific Chemicals A1771936
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| ∼200°C (decomposition) | |
| 19285-83-7 | |
| MFCD00150703 | |
| 14,4469 | |
| OZDAOHVKBFBBMZ-UHFFFAOYSA-N | |
| 1.46 | |
| 500 g | |
| 16219385 | |
| 99% |
| DL-Glutamic acid monohydrate | |
| C5H11NO5 | |
| 4155097 | |
| dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, dl-glutamic acid hydrate, glutaminsaeure hydrate, +/--2-aminoglutaric acid, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, hydrate, hydrate | |
| C(CC(=O)O)C(C(=O)O)N.O | |
| 2-aminopentanedioic acid;hydrate | |
| 165.145 | |
| 165.15 (147.13 Anhydrous) |
Chemical Identifiers
| 19285-83-7 | |
| 165.145 | |
| OZDAOHVKBFBBMZ-UHFFFAOYSA-N | |
| 16219385 | |
| C(CC(=O)O)C(C(=O)O)N.O |
| C5H11NO5 | |
| MFCD00150703 | |
| dl-glutamic acid monohydrate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, dl-glutamic acid hydrate, glutaminsaeure hydrate, +/--2-aminoglutaric acid, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-azanylpentanedioic acid hydrate, hydrate, hydrate | |
| 2-aminopentanedioic acid;hydrate |
Safety and Handling
EINECSNumber : 210-522-2
RTECSNumber : LZ9648000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only