missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethionine 98.0+%, TCI America™

Catalog No. p-7128799
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
E00385G 5 g
E003825G 25 g
2 options

Catalog No. E00385G

Supplier: TCI America E00385G

Only null left

Chemical Identifiers

CAS 67-21-0
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.24
MDL Number MFCD00063102
InChI Key GGLZPLKKBSSKCX-UHFFFAOYNA-N
Synonym dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
PubChem CID 6205
ChEBI CHEBI:68662
IUPAC Name 2-amino-4-(ethylsulfanyl)butanoic acid
SMILES CCSCCC(N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 163.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Ethionine
RTECSNumber ES6825200
TSCA Yes