accessibility menu, dialog, popup

UserName

CAS RN 67-21-0

IUPAC Name: 2-amino-4-(ethylsulfanyl)butanoic acid
Synonyms: (D)-ethionine ethionine DL-
Molecular Weight (g/mol): 163.24
Molecular Formula: C6H13NO2S
InChi Key: GGLZPLKKBSSKCX-UHFFFAOYNA-N
SMILES: CCSCCC(N)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)