missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethionine 98.0+%, TCI America™

Catalog No. E00385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00385G 5 g
E003825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00385G Supplier TCI America Supplier No. E00385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-21-0
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.24
MDL Number MFCD00063102
InChI Key GGLZPLKKBSSKCX-UHFFFAOYNA-N
Synonym dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
PubChem CID 6205
ChEBI CHEBI:68662
IUPAC Name 2-amino-4-(ethylsulfanyl)butanoic acid
SMILES CCSCCC(N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 163.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Ethionine
RTECSNumber ES6825200
TSCA Yes