missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethionine, 98%

CAS: 67-21-0 | C6H13NO2S | 163.24 g/mol

$144.39 - $144.39

Chemical Identifiers

CAS 67-21-0
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.24
MDL Number MFCD00063102
InChI Key GGLZPLKKBSSKCX-UHFFFAOYNA-N
Synonym dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
PubChem CID 6205
ChEBI CHEBI:68662
IUPAC Name 2-amino-4-ethylsulfanylbutanoic acid
SMILES CCSCCC(N)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC146170025
SDS View Documents
Thermo Scientific Chemicals
146170025
2.5 g Glass Bottle
Each for $144.39
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-21-0
163.24
GGLZPLKKBSSKCX-UHFFFAOYNA-N
6205
2-amino-4-ethylsulfanylbutanoic acid
C6H13NO2S
MFCD00063102
dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
CHEBI:68662
CCSCCC(N)C(O)=O

Specifications

265°C to 269°C
White
98%
Glass Bottle
MFCD00063102
dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
CCSCCC(N)C(O)=O
2.5 g
6205
163.23
Crystalline Powder or Flakes
67-21-0
Authentic
C6H13NO2S
C2H5SCH2CH2CH(NH2)CO2H
15, 3792
GGLZPLKKBSSKCX-UHFFFAOYNA-N
2-amino-4-ethylsulfanylbutanoic acid
163.24
CHEBI:68662
≥97.5%
DL-Ethionine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

RUO – Research Use Only