missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine hydrochloride, 90+%

CAS: 10318-18-0 | C3H8ClNO2S | 157.61 g/mol

$210.07 - $1594.07

Chemical Identifiers

CAS 10318-18-0
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064552,MFCD00012632
InChI Key IFQSXNOEEPCSLW-UHFFFAOYNA-N
Synonym dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
PubChem CID 25150
IUPAC Name 2-amino-3-sulfanylpropanoic acid;hydrochloride
SMILES [H+].[Cl-].NC(CS)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2577906
View Documents
Thermo Scientific Chemicals
H2577906
5 g
Each for $210.07
Only null left
 
AAH2577922
View Documents
Thermo Scientific Chemicals
H2577922
100 g
Each for $1,594.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10318-18-0
157.61
IFQSXNOEEPCSLW-UHFFFAOYNA-N
25150
[H+].[Cl-].NC(CS)C(O)=O
C3H8ClNO2S
MFCD00064552,MFCD00012632
dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
2-amino-3-sulfanylpropanoic acid;hydrochloride

Specifications

140°C to 142°C
C3H8ClNO2S
4150473
IFQSXNOEEPCSLW-UHFFFAOYNA-N
2-amino-3-sulfanylpropanoic acid;hydrochloride
157.61
157.62
≥98%
10318-18-0
MFCD00064552,MFCD00012632
dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
[H+].[Cl-].NC(CS)C(O)=O
5 g
25150
Air sensitive; Hygroscopic
DL-Cysteine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 233-698-2

RTECSNumber : HA2200000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only