Learn More
DL-Cysteine hydrochloride, 90+%
CAS: 10318-18-0 | C3H8ClNO2S | 157.61 g/mol
Supplier: Thermo Scientific Chemicals H2577922
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 140°C to 142°C | |
| 10318-18-0 | |
| MFCD00064552,MFCD00012632 | |
| dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride | |
| [H+].[Cl-].NC(CS)C(O)=O | |
| 100 g | |
| 25150 | |
| Air sensitive; Hygroscopic |
| DL-Cysteine hydrochloride | |
| C3H8ClNO2S | |
| 4150473 | |
| IFQSXNOEEPCSLW-UHFFFAOYNA-N | |
| 2-amino-3-sulfanylpropanoic acid;hydrochloride | |
| 157.61 | |
| 157.62 | |
| ≥98% |
Chemical Identifiers
| 10318-18-0 | |
| 157.61 | |
| IFQSXNOEEPCSLW-UHFFFAOYNA-N | |
| 25150 | |
| [H+].[Cl-].NC(CS)C(O)=O |
| C3H8ClNO2S | |
| MFCD00064552,MFCD00012632 | |
| dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride | |
| 2-amino-3-sulfanylpropanoic acid;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 233-698-2
RTECSNumber : HA2200000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only