missing translation for 'onlineSavingsMsg'
Learn More

DL-Camphor, 96%

CAS: 76-22-2 | C10H16O | 152.24 g/mol

Supplier:  Thermo Scientific Chemicals 164640051

Quantity 5 kg
Packaging Plastic drum

 View more versions of this product

Catalog No. AC164640051


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

76-22-2
152.24
DSSYKIVIOFKYAU-UHFFFAOYNA-N
2537
4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
C10H16O
MFCD00074738,MFCD00064149
camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
CHEBI:36773
CC1(C)C2CCC1(C)C(=O)C2

Specifications

DL-Camphor
175°C to 177°C
204°C
95% min. (GC)
C10H16O
MFCD00074738,MFCD00064149
15,1734
Solubility in water: 1.5g/L (20°C). Other solubilities: at 25°C 1 g dissolves in 1 mL alcohol,1 mL ether,,0.5 mL chloroform,0.4 mL benzene,0.4 mL acetone,,1.5 mL turpentine oil,0.5 mL glacial acetic acid.,soluble in aniline,nitrobenzene,c
CC1(C)C2CCC1(C)C(=O)C2
152.24
CHEBI:36773
96%
76-22-2
64g/mL
64°C
Plastic drum
5 kg
07,IV,213
camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
DSSYKIVIOFKYAU-UHFFFAOYNA-N
4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
2537
152.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable solid.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 200-945-

SDS
Documents

Documents

Promotions

Promotions