missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Aspartic acid, 99+%
CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol
$72.48 - $185.48
Chemical Identifiers
| CAS | 617-45-8 |
|---|---|
| Molecular Formula | C4H7NO4 |
| Molecular Weight (g/mol) | 133.10 |
| MDL Number | MFCD00063083 |
| InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
| Synonym | dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart |
| PubChem CID | 424 |
| ChEBI | CHEBI:22660 |
| IUPAC Name | 2-aminobutanedioic acid |
| SMILES | NC(CC(O)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC105032500
|
N/A | 250 g | Plastic Bottle |
Each for $72.48
|
|
||||
|
AC105030010
|
N/A | 1 kg | Plastic Bottle |
Each for $185.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C4H7NO4 | |
| MFCD00063083 | |
| dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
| CHEBI:22660 | |
| NC(CC(O)=O)C(O)=O |
Specifications
| 617-45-8 | |
| Yellow | |
| Authentic | |
| 0.3% max. (105°C, 3 hrs) | |
| C4H7NO4 | |
| MFCD00063083 | |
| −0.3° to +0.3° (20°C, 589nm) (c=10, 2 N HCl) | |
| 10, 853 | |
| dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| 2-aminobutanedioic acid | |
| 424 | |
| 133.1 | |
| Crystalline Low Melting Mass |
| 500ppm max. | |
| 10ppm max. | |
| 10ppm max. | |
| 99+% | |
| HO2CCH2CH(NH2)CO2H | |
| 04, 483 | |
| 500ppm max. | |
| 95% min. (c=10, 2 N HCl, 430nm) 1cm cell | |
| 0.1% max. | |
| NC(CC(O)=O)C(O)=O | |
| 133.10 | |
| CHEBI:22660 | |
| 99.0 to 101.0% (Acid) | |
| DL-Aspartic acid, 99+% |
Safety and Handling
EINECSNumber : 210-513-3
RTECSNumber : CI9097800
TSCA : TSCA
RUO – Research Use Only