missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Aspartic acid, 99+%
CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol
Supplier: Thermo Scientific Chemicals 105030010
| Quantity | 1 kg |
|---|---|
| Packaging | Plastic Bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C4H7NO4 | |
| MFCD00063083 | |
| dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
| CHEBI:22660 | |
| NC(CC(O)=O)C(O)=O |
Specifications
| DL-Aspartic acid | |
| 500ppm max. | |
| Yellow | |
| Authentic | |
| 99+% | |
| Plastic Bottle | |
| HO2CCH2CH(NH2)CO2H | |
| 04, 483 | |
| 500ppm max. | |
| 95% min. (c=10, 2 N HCl, 430nm) 1cm cell | |
| 0.1% max. | |
| NC(CC(O)=O)C(O)=O | |
| 1 kg | |
| 424 | |
| 133.1 | |
| Crystalline Low Melting Mass |
| 99+% | |
| 617-45-8 | |
| 10ppm max. | |
| 10ppm max. | |
| 0.3% max. (105°C, 3 hrs) | |
| C4H7NO4 | |
| MFCD00063083 | |
| −0.3° to +0.3° (20°C, 589nm) (c=10, 2 N HCl) | |
| 10, 853 | |
| dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| 2-aminobutanedioic acid | |
| 133.10 | |
| CHEBI:22660 | |
| 99.0 to 101.0% (Acid) |
Safety and Handling
EINECSNumber : 210-513-3
RTECSNumber : CI9097800
TSCA : TSCA