missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Aspartic acid, 98+%

CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol

Supplier:  Thermo Scientific Chemicals A136460E

 View more versions of this product

Catalog No. AAA136460E


Only null left
Explore available promotions

Description

Description

DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.

Solubility
Soluble in water, and hydrochloric acid. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

>300°C (decomposition)
617-45-8
MFCD00063083
dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
CKLJMWTZIZZHCS-UHFFFAOYNA-N
2-aminobutanedioic acid
133.10
CHEBI:22660
≥98%
DL-Aspartic acid
C4H7NO4
774618
Soluble in water,and hydrochloric acid. Insoluble in ether,and alcohol.
NC(CC(O)=O)C(O)=O
2500 g
424
133.1

Chemical Identifiers

617-45-8
133.10
CKLJMWTZIZZHCS-UHFFFAOYNA-N
424
2-aminobutanedioic acid
C4H7NO4
MFCD00063083
dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart
CHEBI:22660
NC(CC(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-513-3

RTECSNumber : CI9097800

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only