missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%
CAS: 3130-87-8 | C4H8N2O3 | 132.12 g/mol
Supplier: Thermo Scientific Chemicals 167120250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| DL-Asparagine monohydrate | |
| 3130-87-8 | |
| Authentic | |
| C4H8N2O3 | |
| H2NCOCH2CH(NH2)CO2H·H2O | |
| 04, 484 | |
| 14, 837 | |
| 10.5 to 13.5% (K.F.) | |
| NC(CC(N)=O)C(O)=O | |
| 132.12 | |
| 522362 | |
| ≥97.5% |
| 98% | |
| White | |
| 98% | |
| Plastic Bottle | |
| MFCD00151039,MFCD00008036,MFCD00064400 | |
| 90% min. (c=10, 2N HCl, 430nm) 1cm cell | |
| dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina | |
| DCXYFEDJOCDNAF-UHFFFAOYNA-N | |
| 2,4-diamino-4-oxobutanoic acid;hydrate | |
| 25 g | |
| 150.14 | |
| Fine Crystalline Powder |
Chemical Identifiers
| 3130-87-8 | |
| 132.12 | |
| DCXYFEDJOCDNAF-UHFFFAOYNA-N | |
| 522362 | |
| NC(CC(N)=O)C(O)=O |
| C4H8N2O3 | |
| MFCD00151039,MFCD00008036,MFCD00064400 | |
| dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina | |
| 2,4-diamino-4-oxobutanoic acid;hydrate |
Safety and Handling
EINECSNumber : 221-521-1
TSCA : TSCA
RUO – Research Use Only