missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%

CAS: 3130-87-8 | C4H8N2O3 | 132.12 g/mol

$118.86 - $118.86

Chemical Identifiers

CAS 3130-87-8
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00151039,MFCD00008036,MFCD00064400
InChI Key DCXYFEDJOCDNAF-UHFFFAOYNA-N
Synonym dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
PubChem CID 522362
IUPAC Name 2,4-diamino-4-oxobutanoic acid;hydrate
SMILES NC(CC(N)=O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC167120250
SDS View Documents
Thermo Scientific Chemicals
167120250
25 g
Each for $118.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3130-87-8
132.12
DCXYFEDJOCDNAF-UHFFFAOYNA-N
522362
NC(CC(N)=O)C(O)=O
C4H8N2O3
MFCD00151039,MFCD00008036,MFCD00064400
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
2,4-diamino-4-oxobutanoic acid;hydrate

Specifications

3130-87-8
Authentic
C4H8N2O3
H2NCOCH2CH(NH2)CO2H·H2O
04, 484
14, 837
10.5 to 13.5% (K.F.)
NC(CC(N)=O)C(O)=O
132.12
522362
≥97.5%
DL-Asparagine monohydrate, 98%
White
98%
Plastic Bottle
MFCD00151039,MFCD00008036,MFCD00064400
90% min. (c=10, 2N HCl, 430nm) 1cm cell
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
DCXYFEDJOCDNAF-UHFFFAOYNA-N
2,4-diamino-4-oxobutanoic acid;hydrate
25 g
150.14
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-521-1

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only