missing translation for 'onlineSavingsMsg'
Learn More

DL-(+/-)-Arginine Hydrochloride 97.0+%, TCI America™

Catalog No. A05271G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05271G 1 g
A05275G 5 g
A052725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A05271G Supplier TCI America Supplier No. A05271G

Chemical Identifiers

CAS 32042-43-6
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00064549
InChI Key KWTQSFXGGICVPE-UHFFFAOYSA-N
Synonym dl-arginine hydrochloride, 2-amino-5-guanidinopentanoic acid hydrochloride, argininhydrochlorid, dl-arginine hcl, h-dl-arg-oh.xhcl, dl-+/--arginine hydrochloride, 1-hydrochloride arginine, arginine monochloride, h-dl-arg-oh.hcl, wln: muyzm3yzvq &gh
PubChem CID 85880
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 210.66
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-(+/-)-Arginine Hydrochloride