accessibility menu, dialog, popup

UserName

CAS RN 32042-43-6

IUPAC Name: 2-amino-5-[(diaminomethylidene)amino]pentanoic acid hydrochloride
Synonyms: 2-amino-5-[(diaminomethylidene)amino]pentanoic acid hydrochloride
Molecular Weight (g/mol): 210.66
Molecular Formula: C6H15ClN4O2
InChi Key: KWTQSFXGGICVPE-UHFFFAOYNA-N
SMILES: Cl.NC(CCCN=C(N)N)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)