missing translation for 'onlineSavingsMsg'
Learn More

DL-Arginine hydrochloride, 98%, anhydrous, Thermo Scientific Chemicals

Catalog No. p-4323539
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446020050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC446020050 Supplier Thermo Scientific Chemicals Supplier No. 446020050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32042-43-6 | Molecular Formula: C6H14N4O2·ClH | Molecular Weight: 210.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32042-43-6
Molecular Formula C6H14N4O2·ClH
Molecular Weight (g/mol) 210.67
InChI Key KWTQSFXGGICVPE-UHFFFAOYSA-N
Synonym dl-arginine hydrochloride, 2-amino-5-guanidinopentanoic acid hydrochloride, argininhydrochlorid, dl-arginine hcl, h-dl-arg-oh.xhcl, dl-+/--arginine hydrochloride, 1-hydrochloride arginine, arginine monochloride, h-dl-arg-oh.hcl, wln: muyzm3yzvq &gh
PubChem CID 85880
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 200.0°C to 203.0°C
Color White
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5%
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 210.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-Arginine hydrochloride
Show More Show Less
EINECSNumber 250-903-

RUO – Research Use Only