missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Arginine hydrochloride

Catalog No. AAJ6142006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6142006 5 g
AAJ6142014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6142006 Supplier Thermo Scientific Chemicals Supplier No. J6142006

Substrate for nitric oxide synthase; induces insulin release by a nitric oxide-dependent mechanism | CAS: 32042-43-6 | C6H15ClN4O2 | 210.662 g/mol

Substrate for nitric oxide synthase; induces insulin release by a nitric oxide-dependent mechanism

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32042-43-6
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00064549
InChI Key KWTQSFXGGICVPE-UHFFFAOYSA-N
Synonym dl-arginine hydrochloride, 2-amino-5-guanidinopentanoic acid hydrochloride, argininhydrochlorid, dl-arginine hcl, h-dl-arg-oh.xhcl, dl-+/--arginine hydrochloride, 1-hydrochloride arginine, arginine monochloride, h-dl-arg-oh.hcl, wln: muyzm3yzvq &gh
PubChem CID 85880
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 210.67
Physical Form Powder
Chemical Name or Material DL-Arginine hydrochloride
EINECSNumber 250-903-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only