missing translation for 'onlineSavingsMsg'
Learn More

DL-Arabinose 98.0+%, TCI America™

Supplier:  TCI America A05145G

 View more versions of this product

Catalog No. A05145G


Only null left

Description

Description

Specifications

Specifications

DL-Arabinose
158°C
C5H10O5
5 g
SRBFZHDQGSBBOR-UHFFFAOYNA-N
oxane-2,3,4,5-tetrol
5460291
150.13
Crystalline Powder
147-81-9
White
MFCD00135866,MFCD00135608,MFCD00135867,MFCD00135866
l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
OC1COC(O)C(O)C1O
150.13
CHEBI:6182
≥98.0% (HPLC)

Chemical Identifiers

147-81-9
150.13
SRBFZHDQGSBBOR-UHFFFAOYNA-N
5460291
oxane-2,3,4,5-tetrol
C5H10O5
MFCD00135866,MFCD00135608,MFCD00135867,MFCD00135866
l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
CHEBI:6182
OC1COC(O)C(O)C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions